Please login to your online account to display your discounted pricing

EMD Millipore SNAP i.d. 2.0™ Protein Detection Systems


For Immunohistochemistry (Tissue), Immunohistochemistry, Immunohistochemistry (Paraffin)

Manufacturer: emd millipore  SNAP2IHC2

 View more versions of this product

Catalog No. SNAP2IHC2

  • / Each
 

Description & Specifications

Specifications

Storage Conditions Room Temperature
For Use With (Application) Immunohistochemistry (Tissue), Immunohistochemistry, Immunohistochemistry (Paraffin)
Detection Method Chemiluminescence Chromogenic Fluorescence
Includes Base, IHC frame and incubation cover, Assembly fixture, Vacuum tubing and Quick start guide
Quantity 1/Pk.