Please login to your online account to display your discounted pricing

EMD Millipore SNAP i.d.™ 2.0 Protein Detection System


Produces blots of a very high quality every time

Manufacturer: emd millipore  SNAP2MIDI

 View more versions of this product

Catalog No. SNAP2MIDI

  • / Each
 

Description & Specifications

Specifications

Product Type Protein detection system mini
Length (English) Gel 5.31 in.
Width (English) Gel 3.34 in.
Length (Metric) Gel 13.5cm
Width (Metric) Gel 8.5cm
Includes Detection base, 2 midi blot holding frames, 2 antibody collection trays, vacuum tubing, rolling pad and blot roller, 2 wetting trays and a quick start guide
Temperature (Metric) Storage Room Temperature