Please login to your online account to display your discounted pricing
EMD Millipore SNAP i.d.™ 2.0 Protein Detection System: Mini and Midi Blot Holding Frames
Mini and midi blot holding frames for EMD Millipore SNAP i.d. 2.0 Protein Detection Systems
Manufacturer: emd millipore  SNAP2FRMN01
 View more versions of this product
Catalog No. SNAP2FRMN01
  • / Pack
Description & Specifications
Specifications
Product Type Mini Blot Holding Frames
For Use With (Equipment) Snap i.d.™ 2.0 Protein detection system-mini
Includes Clear lids, easy locking latch, umbrella valve
Stackable Yes
Quantity 1/Pk.