Please login to your online account to display your discounted pricing
EMD Millipore SmartPak™ Purification Cartridges
Use for pretreatment of tap water
Manufacturer: emd millipore  SPR00S001US
 View all options for this product
Catalog No. SPR00S001US
Description & Specifications
Model SmartPak™
Product Type RO Purification Pack
For Use With (Equipment) RiOs™ 3 Systems
  • Filtration tecnology used: Activated carbon, reverse osmosis, and ion-exchange