Please login to your online account to display your discounted pricing
EMD Millipore SmartPak™ Purification Cartridges
Use for pretreatment of tap water
$490.00 - $755.00
Specifications
Product Type Low Temperature RO Purification Pack
View More Specs
Products
Catalog Number Mfr. No. Model For Use With (Equipment) Price Quantity    
SPR00S001US
emd millipore
SPR00S001US
SmartPak™ RiOs™ 3 Systems Each for $490.00
SPR0LSIA1US
emd millipore
SPR0LSIA1US
SmartPak™ DQ3 Direct-Q™ 3 and 5 systems Each for $732.00
SPR0LS001US
emd millipore
SPR0LS001US
SmartPak™ RiOs™ 3 and 5 systems Each for $490.00
SPR00SIA1US
emd millipore
SPR00SIA1US
SmartPak™ DQ3 Direct-Q™ 3 systems Each for $755.00
Description & Specifications
Specifications
Product Type Low Temperature RO Purification Pack
  • Filtration tecnology used: Activated carbon, reverse osmosis, and ion-exchange