Please login to your online account to display your discounted pricing

EMD Millipore Simplicity™ Water Purification System


Millipore Simplicity™ Water Purification System provides an economical, efficient, space saving alternative to reagent grade and bottled water.

Manufacturer: emd millipore  SIMS000US

 View more versions of this product

Catalog No. SIMS000US

  • / Each
 

Description & Specifications

Specifications

Particle Size Greater than0.22μm, Less than1 Particulate/mL
Weight (English) 10.8 lb.
Weight (Metric) 4.9kg
Product Type Water purification system
Voltage 100/230V
Height (English) 20.1 in.
Depth (Metric) 36cm
Height (Metric) 51cm
Width (English) 11.4 in.
Contaminant Rejection (Microbiological) Greater than99%
Depth (English) 14.2 in.
Width (Metric) 29cm
Model Simplicity™
Endotoxin Concentration Less than0.001
Flow Rate Greater than0.5
Electrical Requirements 100/230V 50/60Hz
Resistivity 18.2
Product Water Type Type 1
Bacterial Content Less than0.1cfu/mL
Total Organic Carbon Less than15
Certifications/Compliance UL, cUL, CE, ISO 9001, ISO 14001
Hertz 50/60Hz