Please login to your online account to display your discounted pricing

EMD Millipore Simplicity™ Water Purification System

Millipore Simplicity™ Water Purification System provides an economical, efficient, space saving alternative to reagent grade and bottled water.

$3,961.00 - $4,507.00

Specifications

Certifications/Compliance UL, cUL, CE, ISO 9001, ISO 14001
Depth (English) 14.2 in.
Depth (Metric) 36cm
Bacterial Content Less than0.1cfu/mL
Endotoxin Concentration Less than0.001
View More Specs
Products
Catalog Number Mfr. No. Weight (English) Weight (Metric) Total Organic Carbon Price Quantity    

SIMS000US

 
emd millipore
SIMS000US
10.8 lb. 4.9kg Less than15 Each for $3,961.00

SIMSV00US

 
emd millipore
SIMSV00US
11.9 lb. 5.4kg Less than5 Each for $4,507.00
Description & Specifications

Specifications

Certifications/Compliance UL, cUL, CE, ISO 9001, ISO 14001
Depth (English) 14.2 in.
Depth (Metric) 36cm
Bacterial Content Less than0.1cfu/mL
Endotoxin Concentration Less than0.001
Flow Rate Greater than0.5
Product Type Water purification system
Product Water Type Type 1
Resistivity 18.2
Electrical Requirements 100/230V 50/60Hz
Height (English) 20.1 in.
Height (Metric) 51cm
Hertz 50/60Hz
Model Simplicity™
Particle Size Greater than0.22μm, Less than1 Particulate/mL
Voltage 100/230V
Width (English) 11.4 in.
Width (Metric) 29cm

  • Ideal for labs that require 5 to 10L/day ultrapure water
  • System draws water from a removable, refillable built in reservoir
  • Simplicity UV incorporates a dual wavelength UV lamp to reduce TOC and bacteria
  • System Flow Rate: 0.75L/min.