MilliporeSigma™ SeQuant™ ZIC™-HILIC Semi-Preparative Columns
threaded stainless steel HPLC column for semi-preparative HPLC
Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) | Material | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) | Material | Price | Quantity | |||
M1504960001
|
MilliporeSigma™
1504960001 |
20 mm | 50 mm | Stainless Steel |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
SeQuant™ ZIC™-HILIC is the ideal choice for separation of polar and hydrophilic compounds. The high-performance, truly zwitterionic stationary phase, ensures reproductible retention of all types of compounds that have poor retention and are difficult to separate on reversed-phase HPLC columns. ZIC™-HILIC is designed for compounds such as acids and bases, anions and cations, carbohydrates, metabolites, metal complexes, amino acids, peptides, protein digests and much more. This semi-preparative column enables truly scalable transitions from analysis to production.Specifications
HPLC Column |