MilliporeSigma™ SeQuant™ ZIC™-cHILIC Capillary Guard HPLC Column
capillary guard column (3 pc) SeQuant™
$2,231.26 - $2,519.63
Specifications
Length (Metric) 5 mm
Quantity 1/Pk.
View More Specs
Products 2
Catalog Number Mfr. No. Diameter (Metric) Type Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Type Price Quantity  
M1506660001
View Documents
MilliporeSigma™
1506660001
1 mm Guard HPLC Column
Each for $2,231.26
M1506650001
View Documents
MilliporeSigma™
1506650001
0.3 mm HPLC Column
Each for $2,519.63
Description
Description
SeQuant™ ZIC™-cHILIC is designed for complementary selectivity and improved separation of polar hydrophilic compounds. This guard column protects your capillary separation column from contamination.
Specifications
Specifications
5 mm
1/Pk.
SDS