EMD Millipore SeQuant™ ZIC™-cHILIC Capillary Guard HPLC Column
capillary guard column (3 pc) SeQuant®
Manufacturer:  EMD Millipore 1506650001
 View more versions of this product
Catalog No. M1506650001
Specifications
Description
Column, Capillary Guard HPLC; EMD Millipore; SeQuant; ZIC-cHILIC; For separation of polar hydrophilic compounds; I.D.: 0.3mm; Length: 5mm; 1/Pk.
SeQuant® ZIC®-cHILIC is designed for complementary selectivity and improved separation of polar hydrophilic compounds. This guard column protects your capillary separation column from contamination.
Description & Specifications
Specifications
0.3mm
1/Pk.
HPLC Column
5mm