EMD Millipore SeQuant™ SAMS™ and CARS™ Suppressor System: Suppressors
with chemically regenerated membrane (200 cm) for anion chromatography SeQuant®
$6,670.74 - $9,336.94
Specifications
For Use With (Equipment) SeQuant SAMS and CARS Suppressor System
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
M1506090001 EMD Millipore
1506090001
Standard Suppressor Each for $6,670.74
M1506100001 EMD Millipore
1506110001
Gradient Suppressor Each for $9,336.94
Specifications
Description
SeQuant™ CARS™ (Continuous Anion Regeneration System) with SAMS™ (SeQuant™ Anion Membrane Suppressor) will enhance and simplify your anion chromatography analysis. The continuous chemical suppression of the eluent reduces the background and increases the signal for the analyte anions.
This SAMS™ suppressor is suitable for gradients and high ion-strength eluents.
Description & Specifications
Specifications
SeQuant SAMS and CARS Suppressor System