EMD Millipore Samplicity™ Filtration System
Samplicity filtration system with Millex Samplicity membrate filters
$1,097.00 - $1,128.00
Specifications
Material PTFE
Product Type Samplicity™ Filtration System
For Use With (Application) Sample preparation for HPLC, UHPLC, LC/MS, and dissolution testing
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Color Includes Price Quantity  
Catalog Number Mfr. No. Color Includes Price Quantity  
SAMPLG0GR EMD Millipore
SAMPLG0GR
Green 0.2μm Philic PTFE Filters Each for $1,097.00
SAMPLCRGR Millipore Canada Ltd
SAMPLCRGR
Green 0.45um Philic PTFE Filters Each for $1,097.00
SAMPLCRBL Millipore Canada Ltd
SAMPLCRBL
Blue 0.45um Philic PTFE Filters Each for $1,128.00
Specifications
Description
Advantages of the Samplicity filtration system include:
  • Fast simultaneous vacuum filtration of up to eight samples
  • Ergonomic alternative to syringe filters
  • Easy sample preparation of even highly viscous or particulate-laden mixtures
  • Millex Samplicity membrane filters provide long-trusted quality with low extractables and low analyte binding
Ideal for:
  • Dissolution testing
    • High-pressure liquid chromatography (HPLC)
    • Ultra-high performance liquid chromatography (UHPLC or UPLC™)
    • Liquid chromatography – mass spectrometry (LCMS)
Description & Specifications
Specifications
PTFE
Samplicity™ Filtration System
Sample preparation for HPLC, UHPLC, LC/MS, and dissolution testing