Please login to your online account to display your discounted pricing

Samplicity™ Filtration System


Samplicity filtration system with Millex Samplicity membrate filters

Manufacturer: millipore canada ltd  SAMPLCRBL

 View more versions of this product

Catalog No. SAMPLCRBL

  • / Each
 

Description & Specifications

Specifications

Material PTFE
Color Blue
For Use With (Application) Sample preparation for HPLC, UHPLC, LC/MS, and dissolution testing
Includes 0.45um Philic PTFE Filters
Pore Size 0.45μm
Product Type Samplicity™ Filtration System