missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ S-Pak™ Sterile Membrane Filter Kit

Catalog No. HAWG647SP
Change view
Click to view available options
Quantity:
600/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HAWG647SP 600/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. HAWG647SP Supplier MilliporeSigma™ Supplier No. HAWG647SP
Only null left

Made from mixed esters of cellulose and have been optimized for MF method microbiological analysis of water or other liquids

  • Individually packed
  • Gridded membrane filters

  • Beer
  • Bottled Water
  • Cider
  • Clinical Lab
  • Cosmetics
  • Environmental Analysis
  • Food and Beverage
  • Industrial QC
  • Laboratory Filtration
  • Soft Drink
  • Sport Drink
  • Wine

Certifications

Certificates of quality confirm Standard Methods compliance.

Specifications

Color White
Pore Size 0.5 μm
Sterility Sterile
For Use With (Application) For microbiological analysis
Grid Color White
Includes 3 Dispensers
Quantity 600/Pk.
Surface Type Gridded
Diameter (Metric) 47 mm