missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ RiOs™ 3, 5, 8 Pure Water Purification System
Unique, all-in-one purification cartridges include pretreatment and high-performance reverse osmosis (RO) membranes that generate reliable, affordable laboratory-grade water for Type III applications or as pretreatment to ultrapure systems from normal tap
Supplier: MilliporeSigma™ ZR0S0P3US
Description
Produce flow rates adapted to your daily usage—3, 5, or 8L/hour
- Small footprint, easy-to-install system can be placed wherever needed, on the benchtop or wall
- Produce reliable and affordable Type 3 water from tap
- User-friendly, backlit display provides easily accessible information on system status (conductivity, temperature values, water level in reservoir and alarms) at a glance; concise Quick Reference Guide is handy guide for daily operation
- Convenient reservoir storage: RiOs 3 offers an integrated 6L reservoir, RiOs 5 and RiOs 8 can connect to external 30L or 60L reservoirs
- Single SmartPak™ RO cartridge with integrated RO membrane enables easy and rapid replacement
- Efficient removal of >96% of ionic contaminants and ≥99% of other contaminants (organics, bacteria, particles)
- Simple, low-level self-maintenance—just change a single cartridge once or twice a year
Feedwater for ultrapure water systems (Synergy™ or Milli-Q™ systems), humidifiers, autoclaves or glassware washers; manual cleaning and glassware rinsing; buffer preparation
Order Info
19.7 x 11.4 x 13 in. (50 x 29 x 33 cm). 100-250V 50-60Hz. Tap pressure: 0.5 to 6 bar. Feedwater connection: 0.5 in. Gaz M.
Specifications
Single SmartPak™ reverse osmosis | |
<99% | |
3 | |
RiOs Pure Water Purification System | |
Removes more than 96% ionic contaminants, 99% other contaminants | |
29 cm | |
50 cm | |
33 cm | |
6.9 kg | |
100/250 V |
Greater than96% | |
Tap | |
Type 3 | |
Backlit LCD | |
11.4 in. | |
19.7 in. | |
13 in. | |
15.4 lb. | |
100/250 V 50/60 Hz | |
50/60 Hz |