missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore™ Replacement Rolls for MColorpHast™ Neutralit™ pH Indicator Papers, pH 5.5 to 9.0

Roll (4.8 m) with colour scale pH 5.5 - 6.0 - 6.5 - 7.0 - 7.5 - 8.0 - 8.5 - 9.0 Neutralit™

$136.71 - $158.97

Specifications

pH Range 5.5 to 9
Product Type Replacement Roll
For Use With (Application) For long term storage, since they are protected against external influences
For Use With (Equipment) pH indicator papers, 109564, 109565 Neutralit™
View More Specs

Includes: Replacement rolls do not include color scales.

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M1095640003 View Documents EMD Millipore™
1095640003
3 Rolls Pack of 3 Each for $136.71
M1095690001 View Documents EMD Millipore™
1095690001
3 Rolls Each for $158.97
Description

Description

pH indicator papers supplied in rolls are better suited for longterm storage, since they are protected against external influences. pH indicator papers are manufactured by impregnating high-quality filter paper with indicator or mixed-indicator solutions. Therefore, the paper is dryed, cut and packed into the corresponding sizes.

Order Info

Six rolls of 4.8m each

Specifications

Specifications

5.5 to 9
Replacement Roll
For long term storage, since they are protected against external influences
pH indicator papers, 109564, 109565 Neutralit™