MilliporeSigma™ Replacement Rolls for MColorpHast™ Neutralit™ pH Indicator Papers, pH 5.5 to 9.0
Roll (4.8 m) with colour scale pH 5.5 - 6.0 - 6.5 - 7.0 - 7.5 - 8.0 - 8.5 - 9.0 Neutralit™
Manufacturer:  MilliporeSigma™ 1095690001
 View more versions of this product
Catalog No. M1095690001
Description
Description
pH indicator papers supplied in rolls are better suited for longterm storage, since they are protected against external influences. pH indicator papers are manufactured by impregnating high-quality filter paper with indicator or mixed-indicator solutions. Therefore, the paper is dryed, cut and packed into the corresponding sizes.
Specifications
Specifications
For long term storage, since they are protected against external influences
3 Rolls
5.5 to 9
pH indicator papers, 109564, 109565 Neutralit™
Replacement Roll