EMD Millipore Replacement Receiver Flasks
Provide a large filter area for rapid sterile filtration of aqueous fluids into narrow-mouth (33mm) or wide-mouth (45mm) bottles
$56.50 - $69.50
Specifications
Material PS (Polystyrene)
Sterility Sterile
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
SC00B02RE EMD Millipore
SC00B02RE
250mL Pack of 12 Each for $56.50
SC00B05RE EMD Millipore
SC00B05RE
500mL Pack of 12 Each for $69.50
Specifications
Description
  • Sterile, polystyrene
  • Can be used as receivers for Millipore Steritop™ Vacuum Bottle-Top Filters
  • 45mm neck with screw-top lid
  • Storage of sterilized or clarified fluids
  • Laboratory Filtration
Description & Specifications
Specifications
PS (Polystyrene)
Sterile