Please login to your online account to display your discounted pricing
EMD Millipore Replacement Parts for Swinnex™ Filter Holders
Use to ultraclean or sterilize small volumes of liquids dispensed by syringe
Manufacturer: emd millipore  SX0002501
 View more versions of this product
Catalog No. SX0002501
  • / Pack of 100 Each
Description & Specifications
Specifications
For Use With (Equipment) Millipore Swinnex Filter Holder
Description 25mm Gasket
Material Silicone
Diameter (Metric) Inner 25mm