EMD Millipore Replacement Parts for Swinnex™ Filter Holders
Use to ultraclean or sterilize small volumes of liquids dispensed by syringe
$51.50 - $198.00
Specifications
For Use With (Equipment) Millipore Swinnex Filter Holder
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
SX0004701 EMD Millipore
SX0004701
47mm O-rings Base Pack of 5 Each for $58.00
SX0002501 EMD Millipore
SX0002501
25mm Gasket Pack of 100 Each for $125.00
SX0001301 EMD Millipore
SX0001301
13mm Gasket Pack of 100 Each for $62.50
XX1104707 EMD Millipore
XX1104707
47mm Filter O-ring Pack of 10 Each for $51.50
XX2702510 EMD Millipore
XX2702510
25mm Support Screens Pack of 30 Each for $198.00
Specifications
Description
Connect Swinnex 47mm with tubing to pressurized reservoir or automatic filling machine for filtering larger volumes.
Description & Specifications
Specifications
Millipore Swinnex Filter Holder