Please login to your online account to display your discounted pricing
EMD Millipore Replacement Parts for Stainless-Steel 13 and 25mm Filter Holders
Use to ultraclean or sterilize small volumes of liquids dispensed by syringe
Manufacturer: emd millipore  XX30012RK
 View more versions of this product
Catalog No. XX30012RK
  • / Each
Description & Specifications
Specifications
Description Stainless-Steel Filter Holder Replacement Parts
Includes Two PTFE gaskets, two support screens and O-rings
For Use With (Equipment) Stainless-steel 13mm filter holders
Material Stainless Steel