EMD Millipore Replacement Parts for Stainless-Steel 13 and 25mm Filter Holders
Use to ultraclean or sterilize small volumes of liquids dispensed by syringe
Manufacturer:  EMD Millipore XX3001210
 View more versions of this product
Catalog No. XX3001210
Specifications
Description
Support screen, Stainless steel; EMD Millipore; 1 per pack
Swinnex™ parts kit contains two PTFE gaskets, two O-rings, and two support screens.
Description & Specifications
Specifications
Stainless-Steel Filter Holder Replacement Parts
Stainless Steel
Support screen