Please login to your online account to display your discounted pricing

EMD Millipore Replacement Parts for Stainless-Steel 13 and 25mm Filter Holders


Use to ultraclean or sterilize small volumes of liquids dispensed by syringe

Manufacturer: emd millipore  XX30025RK

 View more versions of this product

Catalog No. XX30025RK

  • / Each
 

Description & Specifications

Specifications

Description Stainless-Steel Filter Holder Replacement Parts
Includes Two PTFE gaskets, two O-rings, and two support screens
For Use With (Equipment) Stainless-steel 13mm filter holders
Material Stainless Steel
Diameter (Metric) Inner 25mm