missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Accessories for Solvent Filtering Dispensers, 25 mm
Accessories for filter holders for sample preparation.
Supplier: MilliporeSigma™ XX3002502
Description
AccessoriesXX6602501 Flask and Neoprene StopperXX3002502 Filter Holder Gasket, PTFEXX3002510 Filter Holder Support Screen, stainless steelSpecifications
Solvent Filtering Dispenser | |
Filter Holder Gasket | |
2.5 cm |
PTFE | |
0.98 in. |