Please login to your online account to display your discounted pricing

EMD Millipore Replacement Membranes with Pads for EMD Millipore Fluid Contamination Analysis Monitors

Has thick absorbent pad for liquid monitoring

$153.00 - $269.58

Specifications

Color White
Sterile No
View More Specs
Products
Catalog Number Mfr. No. For Use With (Application) Pore Size Price Quantity    

AAWP037P0

 
emd millipore
AAWP037P0
Protein purification, analytical sample preparation 0.8μm Pack of 100 Each for $153.00

SMWP03700

 
emd millipore
SMWP03700
Patch test for fluids and air monitoring 5μm Pack of 100 Each for $269.58
Description & Specifications

Specifications

Color White
Sterile No

  • Refill for EMD Millipore™ fluid contamination analysis monitors
  • MF-Millipore MCE membranes for 37mm contamination monitors
  • Composed of mixed cellulose esters with thick cellulose pad
  • For use with EMD Millipore model no. MAWP037P0, MAWP037PM, M000037P0