Please login to your online account to display your discounted pricing

Reflectoquant™ pH Reflectometric Tests with Strips


For use with Reflectoquant™ systems

Manufacturer: emd millipore corporation  1168940001

 View more versions of this product

Catalog No. M1168940001

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Used for monitoring the pH value in aqueous solutions, beverages
pH Range 1 to 5
For Use With (Equipment) Reflectometric
Test Method Mixed indicator
Storage Requirements 15° to 25°C
Quantity 50 Tests
Product Type pH Test