missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Reflectoquant™ Peroxide Test Strips

For determination of inorganic peroxides in aqueous solutions

$244.40 - $4,826.48

Specifications

For Use With (Application) For food and water analysis
Storage Requirements 2° to 8°C
View More Specs

Includes: Tube containing 50 strips, 1 bar-code strip.

Products 4
Catalog Number Mfr. No. Product Type Graduations No. of Tests Price Quantity  
Catalog Number Mfr. No. Product Type Graduations No. of Tests Price Quantity  
M1169740001 View Documents MilliporeSigma™
1169740001
Test Strip 0.2 to 20mg/L 50.0 Each for $255.31
 
M1094500010 View Documents MilliporeSigma™
1094500010
pH Strip 2-2.5-3-3.5-4-4.5-5-5.5-6-6.5-7-7.5-8-8.5-9 1000.0 Each for $776.91
 
M1094500013 View Documents MilliporeSigma™
1094500013
pH Strip 2-2.5-3-3.5-4-4.5-5-5.5-6-6.5-7-7.5-8-8.5-9 25000.0 Each for $4,826.48
 
M1167310001 View Documents MilliporeSigma™
1167310001
Test Strip 100 to 1000mg/L 50.0 Each for $244.40
 
Description

Description

  • Enzymatic reaction method ideal for screening
  • Barcode calibration for reliable, quantitative results
  • Low analysis costs

Reclose the tube containing the test strips immediately after use.

For food and water analysis

Specifications

Specifications

For food and water analysis
2° to 8°C