Please login to your online account to display your discounted pricing

EMD Millipore Reflectoquant™ Peracetic Acid Test Strips


For determination of inorganic peracetic acid in aqueous solutions

Manufacturer: emd millipore  1169760001

 View more versions of this product

Catalog No. M1169760001

  • / Each
 

Description & Specifications

Specifications

No. of Tests 50.0
Method Redox Reaction
Storage Requirements 2° to 8°C
Graduations 75 to 400mg/L
Product Type Test Strip
Range 75 to 400mg/L
For Use With (Application) Food and water analysis, ideal for determining when hydrogen peroxide is present in a sample
For Use With (Equipment) RQflex™ 10 and RQflex™ Plus 10 reflectometers