Please login to your online account to display your discounted pricing

EMD Millipore Reflectoquant™ Iron Test Strips


To determine the presence of residues of iron

Manufacturer: emd millipore  1169830001

 View more versions of this product

Catalog No. M1169830001

  • / Each
 

Description & Specifications

Specifications

No. of Tests 50.0
Method 2.2'-Bipyridine
Product Type Test Strip
Range 20 to 200mg/L
Storage Requirements 15° to 25°C
Graduations 20 to 200mg/L
For Use With (Application) Ideal for measuring iron(II) ions in surface water, seawater, industrial water, wastewater and mineral water
For Use With (Equipment) RQflex™ 10 and RQflex™ Plus 10 reflectometers