Please login to your online account to display your discounted pricing
Quant™ Test Strips
Easy-to-use test strips for field testing, preliminary semi-quantitative analysis or quality control
Manufacturer: emd millipore corporation  1.10020.0001
 View more versions of this product
Catalog No. M100201
  • / Pack of 100 Each
Description & Specifications
Specifications
For Use With (Application) Field testing, preliminary semi-quantitative analysis or quality control
Product Type Test: Nitrate
Range 10 to 500ppm