Please login to your online account to display your discounted pricing
Quant™ Test Strips
Easy-to-use test strips for field testing, preliminary semi-quantitative analysis or quality control
Manufacturer: thermo sci diagnostix  M100031
 View more versions of this product
Catalog No. M100031
  • / Each
Description & Specifications
Specifications
For Use With (Application) Field testing, preliminary semi-quantitative analysis or quality control
Product Type Test: Copper; Range: 10-300ppm