Please login to your online account to display your discounted pricing
EMD Millipore Qualification Workbook
Provides instructions on performing system qual and completing qual forms
Manufacturer: emd millipore  VFRX0X045
 View more versions of this product
Catalog No. VFRX0X045
  • / Each
Description & Specifications
Specifications
For Use With (Application) Provides instructions on performing system qual and completing qual forms
For Use With (Equipment) Milli-RX™ 45 system
Includes IQ and OQ protocols, recommended PQ protocols and annual system maintenance procedures
Product Type Qualification workbook