EMD Millipore Pure Water Storage Reservoir Distribution Pump
Magnetically coupled centrifugal pump delivers pressurized water to water purification systems or laboratory equipment such as glassware washers
$285.00 - $1,272.00
Specifications
For Use With (Equipment) Pump TANK REC 56
View More Specs
Requires: Pump Connection Kit (PUMP KIT 01), which contains 12mm polyethylene tubing, check valve, connectors, elbows and tees.
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
PUMPKIT01 EMD Millipore
PUMPKIT01
12mm Polyethylene tubing, check valve, connectors, elbows and tees Each for $285.00
TANKREC56 EMD Millipore
TANKREC56
12mm Polyethylene tubing, check valve, connectors, elbows, tees and distribution pump Each for $1,272.00
Specifications
Description
  • Fits into base of EMD Millipore™ Pure Water Storage Reservoirs
  • Provides quiet, maintenance-free operation for up to 35,000 hours
  • Delivers water continuously without significantly raising water temperature
Description & Specifications
Specifications
Pump TANK REC 56