Please login to your online account to display your discounted pricing
EMD Millipore Pretreatment Packs
For use in EMD Millipore ELIX and RiOs™ Systems
$602.00 - $602.00
Specifications
Product Type Pretreatment pack
View More Specs
Products
Catalog Number Mfr. No. Purification Method For Use With (Equipment) Price Quantity    
PR0GTL0S1US
emd millipore
PR0GTL0S1US
Activated Carbon, depth filtration, anti-scaling Elix™, RiOs™, Large Elix™, Large RiOs™ Each for $602.00
Description & Specifications
Specifications
Product Type Pretreatment pack