Please login to your online account to display your discounted pricing

EMD Millipore Pretreatment Packs

For use in EMD Millipore ELIX and RiOs™ Systems

Manufacturer: emd millipore  PR0GTL0S1US

 View all options for this product

Catalog No. PR0GTL0S1US


Description & Specifications


Product Type Pretreatment pack
For Use With (Equipment) Elix™, RiOs™, Large Elix™, Large RiOs™
Purification Method Activated Carbon, depth filtration, anti-scaling