Please login to your online account to display your discounted pricing

EMD Millipore Prep/Scale Spiral-Wound Ultrafiltration Modules: TFF-6


Simple design for easy setup in TFF systems

Manufacturer: emd millipore  SK1P003W4

 View more versions of this product

Catalog No. SK1P003W4

  • / Each
 

Description & Specifications

Specifications

Diameter (English) Outside 2.3 in.
Diameter (Metric) Outside 5.8cm
Filter Code PLGC
Molecular Weight 10 NMWL
Quantity 1 Pack
Filtration Area 5400cm2
Format Spiral Wound Cartridge
Length (English) 15.7 in.
Length (Metric) 39.9cm
Volume (Metric) Working 250mL
For Use With (Application) Economical preparation of laboratory scale samples up to small production volumes by tangential flow filtration
Certifications/Compliance USP, Class VI
Filter Material Regenerated Cellulose
Filter Type Ultrafiltration
Material Polyethylene feed/retentate screen, Polypropylene Permeate screen, Polysolfone housing and core with polyurethane adhesive, PTFE with Silicone reinforcement Tape
Max. Inlet Pressure 0 to 80psi
Recirculation Rate 1 to 6L/min.
Volume (English) Working 8.45 oz.
Product Type Prep/Scale Ultrafiltration Module
pH Range 2 to 13