Please login to your online account to display your discounted pricing

EMD Millipore Prep/Scale Spiral-Wound Ultrafiltration Modules: TFF-2


For easy, reliable preparation of samples ranging from 100L down to 100mL

Manufacturer: emd millipore  SK1P026W3

 View more versions of this product

Peristaltic pump, flexible tubing and feed solution

Catalog No. SK1P026W3

  • / Each
 

Description & Specifications

Specifications

Diameter (English) Outside 2.3 in.
Filter Type Ultrafiltration
Filtration Area 2300cm2
Diameter (Metric) Outside 5.8cm
Filter Material Regenerated Cellulose
Format Spiral Wound Cartridge
Length (Metric) 23.4cm
Max. Inlet Pressure 0 to 80psi
pH Range 2 to 13
Certifications/Compliance USP, Class VI
Length (English) 9.2 in.
Material Polyethylene feed/retentate screen, Polypropylene Permeate screen, Polysolfone housing and core with polyurethane adhesive, PTFE with Silicone reinforcement Tape
Molecular Weight 10 NMWL
Recirculation Rate 1 to 6L/min.
Volume (English) Working 5.07 oz.
Volume (Metric) Working 150mL
Quantity 1 Pack
Filter Code PLGC
For Use With (Application) Economical preparation of laboratory scale samples up to small production volumes by tangential flow filtration
Product Type Prep/Scale Ultrafiltration Module