EMD Millipore PetriSlide™ Containers
Use in place of Petri dishes
$79.00 - $79.00
Specifications
Diameter (Metric) 47 mm
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Color For Use With (Application) Price Quantity  
Catalog Number Mfr. No. Color For Use With (Application) Price Quantity  
PD1504700 EMD Millipore
PD1504700
Transparent cover For use without pads for contamination analysis Pack of 100 Each for $79.00
Specifications
Description
Sterilized polystyrene storage and examination containers for culturing microorganisms.
  • Hold 47mm filter securely in place
  • Transparent cover allows microscopic examination without removal
  • Rectangular base has rounded corners for mounting on microscopic stage
  • Stored in cartons or standard slide cases
  • Use pure cellulose pads for microbiological analysis
  • Use without pads for contamination analysis
Description & Specifications
Specifications
47 mm