EMD Millipore™ Petri Dishes and Petri-Pad™ Dishes
Available with or without absorbent pads
Manufacturer:  EMD Millipore™ PD20047S5
 View more versions of this product
Catalog No. PD20047S5
Requires : When using with liquid medium, add sterile absorbent pads (order Cat. No. AP10 047 S0, 47mm Sterile Pads; or AP10 045 S1, Millipore Pad Dispenser with Sterile Pads)
Description
Description
Petri Dishes
  • 47mm sterilized Petri dishes
  • Use with solid agar medium
  • Dishes are made of molded polystyrene
Petri-Pad Dishes
  • Presterilized disposable 47mm dishes preloaded with sterilized absorbent pads
  • Eliminate labor and cost of hand-loading pads and minimize contamination
  • Pads are pure cellulose
Specifications
Specifications
47 mm
Sleeve
Yes
Sterile
600 Pk. (25 sleeves); 20/Sleeve
Dish with pad
Documents