missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore™ Petri Dishes and Petri-Pad™ Dishes

Available with or without absorbent pads

Manufacturer:  EMD Millipore™ PD20047S0

 View more versions of this product

Catalog No. PD20047S0



Requires : When using with liquid medium, add sterile absorbent pads (order Cat. No. AP10 047 S0, 47mm Sterile Pads; or AP10 045 S1, Millipore Pad Dispenser with Sterile Pads)

Description

Description

Petri Dishes

  • 47mm sterilized Petri dishes
  • Use with solid agar medium
  • Dishes are made of molded polystyrene
Petri-Pad Dishes
  • Presterilized disposable 47mm dishes preloaded with sterilized absorbent pads
  • Eliminate labor and cost of hand-loading pads and minimize contamination
  • Pads are pure cellulose

Specifications

Specifications

47 mm
Sleeve
Yes
Sterile
150 Pk.
Dish with pad
Documents