missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Pellicon™ XL Filters, Durapore™ Membranes - Hydrophilic PVDF

Polypropylene cassettes with Durapore membrane

Manufacturer:  MilliporeSigma™ PXGVPPC50

 View more versions of this product

Catalog No. PXGVPPC50



Description

Description

Durapore DVPP
Suitable for harvesting cell cultures or colloidal suspensions

Durapore GVPP
Recommended for harvesting and washing colloidal suspensions, bacterial cells; clarification of protein solutions and viral cultures

Durapore HVMP
Used to harvest and wash colloidal suspensions as well as cell and viral cultures; and to clarify protein solutions and viral cultures

Durapore VVPP
For the clarification of cell lysates, protein solutions and viral cultures

Specifications

Specifications

7.4 x 1.2 in. (18.8 x 3cm)
Microfiltration
Pellicon™ XL device and Labscale™ TFF system, Pellicon™ XL 50 microfiltration cassettes
Hydrophilic
18.8cm
0.03 to 0.05L/min.
3.2mL
1.2 in.
Protein purification, analytical sample preparation
0.22μm
GVPP
50cm2
Cassette
7.4 in.
5.5 bar
0.5 oz.
15mL
3cm
Polypropylene
1.5 to 9 continuous, 1 to 11 cleaning
Documents