Please login to your online account to display your discounted pricing

EMD Millipore Pellicon™ XL Devices


Small-volume tangential flow filter device

Manufacturer: emd millipore  PXB008A50

 View more versions of this product

Catalog No. PXB008A50

  • / Each
 

Description & Specifications

Specifications

Quantity 1 Pack
Product Type Pellicon™ XL Ultrafiltration Device
For Use With (Application) Protein purification, analytical sample preparation
Filtration Area 50cm2
Filter Type Ultrafiltration Biomax-8
Includes Operating instructions, certificate of quality, and accessory kit (four luer barb fittings, 4 tubing clamps, one retentate clamp, 4 ft. of LFL Tygon™ tubing)
Length (English) 7.4 in.
Length (Metric) 18.8cm
Material Polypropylene
Max. Inlet Pressure 80psi at 50°C
Molecular Weight 8kDa
Pore Size 100μm
pH Range 1 to 14
Recirculation Rate 0.03 to 0.05L/min.
Volume (English) Working 0.5 oz.
Volume (Metric) Hold Up 3.2mL
Volume (Metric) Working 15mL
Width (English) 1.2 in.
Width (Metric) 3cm
Format Cassette
Dimensions (L x W) 7.4 x 1.2 in. (18.8 x 3cm)

Designed for process development and sample preparation, with an optimal processing volume range of 50 to 1000mL.

  • True linear scalability with same flow path, channel length, and channel height as other established Pellicon products
  • Can operate with peristaltic pump or Labscale™ TFF system Robust thermoplastic (polypropylene) construction
  • Controlled channel height
  • Choice of Biomax™ PES (polyethersulfone) or Ultracel™ PLC (composite regenerated cellulose) membranes

Concentration and desalting of proteins, buffer exchange, depyrogenation, clarification, and preparation of material for clinical trials.