Please login to your online account to display your discounted pricing

EMD Millipore Packs and Kits for Millipore Compact Water Systems For Milli-Q™ Compact Systems


Combination of purification media selected to remove typical contaminants

Manufacturer: emd millipore  CPMQ004D2

 View more versions of this product

Catalog No. CPMQ004D2

  • / Each
 

Description & Specifications

Specifications

Purification Method Ion exchange, depth filtration
Product Type Purification Pack
For Use With (Equipment) Milli-Q and Milli-RO Purification Systems
Model Quantum IX (Ionex)1
Feedwater Source Deionized
Resistivity 18 megohm-cm