Please login to your online account to display your discounted pricing

EMD Millipore Packs and Kits for Millipore Compact Water Systems For Milli-Q™ Compact Systems


Combination of purification media selected to remove typical contaminants

Manufacturer: emd millipore  CPMQK05D2

 View more versions of this product

Catalog No. CPMQK05D2

  • / Each
 

Description & Specifications

Specifications

Purification Method Ion exchange, depth filtration, membrane filter
For Use With (Equipment) Milli-Q and Milli-RO Purification Systems
Model QPAK 2
Includes Purification pack, Millipak™ final filter
Feedwater Source Deionized water
Pore Size 0.2μm