Please login to your online account to display your discounted pricing

EMD Millipore Packs and Kits for Millipore Compact Water Systems For Milli-Q™ Compact Systems


Combination of purification media selected to remove typical contaminants

Manufacturer: emd millipore  CPMQ004R1

 View more versions of this product

Catalog No. CPMQ004R1

  • / Each
 

Description & Specifications

Specifications

For Use With (Equipment) Milli-Q and Milli-RO Purification Systems
Model Quantum IX (Ionex)1
Purification Method Ion exchange, activated Carbon
Flow Rate 2
Feedwater Source Reverse osmosis, distilled water