missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Optifix™ Bottletop Dispensers
For use with organic solvents and materials
Supplier: MilliporeSigma™ 101107481
Description
- Choice of basic, solvent or safety models
- Autoclavable
Specifications
0.3% | |
Autoclavable | |
Intake tube, certificate of precision | |
±0.1% |
Safety Bottletop Dispenser | |
10 to 50 mL | |
1 mL |