missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Optifix™ Bottletop Dispensers

For use with organic solvents and materials

Supplier:  MilliporeSigma™ 101081511

 View more versions of this product

Catalog No. M101081511



Description

Description

  • Choice of basic, solvent or safety models
  • Autoclavable

Specifications

Specifications

0.3%
Autoclavable
Intake tube, certificate of precision, 38, 40, 45mm polypropylene adapters
±0.1%
Solvent Bottletop Dispenser
20 to 100 mL
2 mL
Lifetime
SDS
Documents

Documents

Promotions

Promotions