missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Optifix™ Bottletop Dispensers

For use with organic solvents and materials

Manufacturer:  MilliporeSigma™ 101081481

 View more versions of this product

Catalog No. M101081481



Description

Description

  • Choice of basic, solvent or safety models
  • Autoclavable

Specifications

Specifications

Autoclavable
Solvent Bottletop Dispenser
1 mL
±0.1%
0.3%
10 to 50 mL
Intake tube, certificate of precision, 38, 40, 45mm polypropylene adapters
Lifetime
Documents
Promotions

Promotions