EMD Millipore Opticap™ XL2 Disposable Capsule Filters with Polysep II™ Media
Ideal for a broad range of aqueous-based applications
$314.00 - $314.00
Specifications
Autoclavable Yes
Material (Membrane) Mixed Cellulose Esters
Filtration Area (Metric) 0.06m2
For Use With (Application) Laboratory Filtration; Buffer Preparation; Cell Culture Media; Large Volume Parenterals (LVP); Ophthalmics; Serum
Length (English) 5.6 in.
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Inlet Style Outlet Style Pore Size Price Quantity  
Catalog Number Mfr. No. Inlet Style Outlet Style Pore Size Price Quantity  
KGW6A02HH3 EMD Millipore
KGW6A02HH3
0.56 in. (14 mm) Hose Barb 0.56 in. (14 mm) Hose Barb 1.0 + 0.5μm Pack of 3 Each for $314.00
KGW9A02HH3 EMD Millipore
KGW9A02HH3
0.56 in. (14 mm) Hose Barb 0.56 in. (14 mm) Hose Barb 2.0 + 1.2μm Pack of 3 Each for $314.00
Specifications
Description
  • Media consists of a borosilicate glass layer and a layer of mixed esters of cellulose membrane
  • Depth of the microfiber filter layer provides high particle loading capacity, and retention of large particulates while maintaining high flow rates
  • Excellent thermal and hydraulic stress resistance
  • Capsule design with pleated Polysep II media minimizes hold-up volume and reduces production losses
Not in-line steam sterilizable
Laboratory Filtration; Buffer Preparation; Cell Culture Media; Large Volume Parenterals (LVP); Ophthalmics; Serum
Order Info
Meets USP Oxidizable Substances Test requirements after a water flush of ≥1L.
Description & Specifications
Specifications
Yes
0.06m2
5.6 in.
No
Mixed Cellulose Esters
Laboratory Filtration; Buffer Preparation; Cell Culture Media; Large Volume Parenterals (LVP); Ophthalmics; Serum
14.2cm