MilliporeSigma™ Opticap™ Sterile XL150 Capsule Filters
For sterilizing grade performance and mycoplasma removal
Supplier:  MilliporeSigma™ KHGES015HH3
 View more versions of this product
Catalog No. KHGES015HH3
Description
Description
  • High-flux hydrophilic sterilizing-grade 0.1μm rated PES membrane for improved process economy
  • On-board PES membrane prefilter for extended throughput in high fouling media
  • Robust construction
  • Withstand multiple sterilization cycles
  • Easy wetting and integrity testing
  • Broad chemical compatibility across a wide pH range (pH 1–14)
  • Hydrophilic Polyethersulfone filter
  • Polyester supports
  • Structural components: Gamma stable polypropylene/Polysulfone
  • Silicone vent O-rings
  • Gamma compatible sterilization
Cell culture media; Media additives; Serum; Concentrated protein; Process intermediates; Biologicals
Specifications
Specifications
0.5/0.2 μm
Cell culture media; Media additives; Serum; Concentrated protein; Process intermediates; Biologicals
0.56 in. (14mm) hose barb
9.7 cm
0.014 m2
0.56 in. (14mm) hose barb
3.8 in.